3-Methyl-1-butene 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Isoamylene, Isopropylethylene, Isopentene
OR460043-100G
OR460043-25G
OR460043-5G
APOSOR460043-100GEA
400
EUR
APOSOR460043-100G
APOSOR460043-25G
APOSOR460043-5G
3-Methyl-1-butene 95%
3-Methyl-1-butene
Formula:
C₅H₁₀ Boiling Pt: 20 °C Melting Pt: –168.5…–168 °C Density: 0.627 g/cm³ (25 °C) Flash Pt: 57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008937 CAS Number: 563-45-1 UN: 2561 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...