1-Bromo-3-methyl-2-butene 90+%
Supplier: Apollo Scientific
Danger
Synonyms: 3,3-Dimethylallyl bromide, 4-Bromo-2-methyl-2-butene
OR46013-50G OR46013-250G
APOSOR46013-50GEA 219 EUR
APOSOR46013-50G APOSOR46013-250G
1-Bromo-3-methyl-2-butene 90+%
1-Bromo-3-methyl-2-butene
Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 82…83 °C/150mmHg
Density: 1.29 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000242
CAS Number: 870-63-3
EINECS: 212-799-5
UN: 2920
ADR: 8+3 II