2-Methoxyphenyl benzoate 99%
Supplier: Apollo Scientific
Warning
OR46021-25G OR46021-100G
APOSOR46021-25GEA 59.6 EUR
APOSOR46021-25G APOSOR46021-100G
2-Methoxyphenyl benzoate 99%
2-Methoxyphenyl benzoate
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 57…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00017153
CAS Number: 531-37-3
EINECS: 208-507-0