3-Indolylglyoxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 1H-Indol-3-yl(oxo)acetic acid, 3-Indoleglyoxylic Acid
OR46044-1G OR46044-5G
APOSOR46044-1GEA 80 EUR
APOSOR46044-1G APOSOR46044-5G
3-Indolylglyoxylic acid
3-Indolylglyoxylic acid
Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 217 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005625
CAS Number: 1477-49-2
EINECS: 216-029-9