Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate 95%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 6-chloro-3,5-diaminopyrazine-2-carboxylate, Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
OR46066-25G OR46066-5G
APOSOR46066-25GEA 63.8 EUR
APOSOR46066-25G APOSOR46066-5G
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate 95%
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate
Formula: C₆H₇ClN₄O₂
MW: 202.60 g/mol
Melting Pt: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD01928388
CAS Number: 1458-01-1
EINECS: 215-946-1