2-(1H-Pyrazol-4-yl)ethanol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(2-Hydroxyethyl)-1H-pyrazole
OR46067-25G OR46067-5G OR46067-1G
APOSOR46067-25GEA 192 EUR
APOSOR46067-25G APOSOR46067-5G APOSOR46067-1G
2-(1H-Pyrazol-4-yl)ethanol 98%
2-(1H-Pyrazol-4-yl)ethanol
Formula: C₅H₈N₂O
Boiling Pt: 140…145 °C/0.3mmHg
Melting Pt: 54…55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03419343
CAS Number: 180207-57-2