4-Bromo-4'-tert-butylbiphenyl
Supplier: Apollo Scientific
Warning
OR46097-5G OR46097-1G
APOSOR46097-5GEA 37.7 EUR
APOSOR46097-5G APOSOR46097-1G
4-Bromo-4'-tert-butylbiphenyl
4-Bromo-4'-tert-butylbiphenyl
Formula: C₁₆H₁₇Br
MW: 289.21 g/mol
Boiling Pt: 152 °C/2mmHg
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD01321141
CAS Number: 162258-89-1