2,6-Dibromonaphthalene 99%
Supplier: Apollo Scientific
Danger
OR46098-1G OR46098-25G OR46098-5G
APOSOR46098-1GEA 26.1 EUR
APOSOR46098-1G APOSOR46098-25G APOSOR46098-5G
2,6-Dibromonaphthalene 99%
2,6-Dibromonaphthalene
Formula: C₁₀H₆Br₂
MW: 285.97 g/mol
Melting Pt: 158…163 °C
Storage Temperature: Ambient
MDL Number: MFCD01026462
CAS Number: 13720-06-4
UN: 3077
ADR: 9+EHS,III