2,6-Dichloro-3-formylpyridine
Supplier: Apollo Scientific
Danger
Synonyms: 2,6-Dichloronicotinaldehyde, 2,6-Dichloropyridine-3-carboxaldehyde, 2,6-Dichloro-3-formylpyridine, 2,6-Dichloro-3-pyridinecarboxaldehyde
OR46107-25G
APOSOR46107-25GEA 59.6 EUR
APOSOR46107-25G
2,6-Dichloro-3-formylpyridine
2,6-Dichloro-3-formylpyridine
Formula: C₆H₃Cl₂NO
MW: 176.0001 g/mol
Boiling Pt: 95 °C/3mmHg
Melting Pt: 74…75 °C
Storage Temperature: Ambient
MDL Number: MFCD07369746
CAS Number: 55304-73-9
UN: 2811
ADR: 6.1,III