(R)-(-)-2-Methylpyrrolidine
Supplier: Apollo Scientific
Danger
Synonyms: (R)-2-Methylpyrrolidine
OR4617-1G OR4617-5G
APOSOR4617-1GEA 164 EUR
APOSOR4617-1G APOSOR4617-5G
(R)-(-)-2-Methylpyrrolidine
(R)-(-)-2-Methylpyrrolidine
Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 97…99 °C/744mmHg
Density: 0.842 g/cm³ (25 °C)
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07783026
CAS Number: 41720-98-3
UN: 2925
ADR: 4.1+8,II