5-Methoxybenzofuran
Supplier: Apollo Scientific
Warning
OR46210-1G OR46210-5G
APOSOR46210-1GEA 58 EUR
APOSOR46210-1G APOSOR46210-5G
5-Methoxybenzofuran
5-Methoxybenzofuran
Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 123 °C/20mmHg
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00079770
CAS Number: 13391-28-1