(R)-2,3-Dihydro-1H-inden-1-amine hydrochloride 95+%
Supplier: Apollo Scientific
Warning
Synonyms: (1R)-(-)-1-Aminoindane hydrochloride
OR46214-100G OR46214-25G OR46214-5G
APOSOR46214-100GEA 876 EUR
APOSOR46214-100G APOSOR46214-25G APOSOR46214-5G
(R)-2,3-Dihydro-1H-inden-1-amine hydrochloride 95+%
(R)-2,3-Dihydro-1H-inden-1-amine hydrochloride
Formula: C₉H₁₂ClN
MW: 169.6513 g/mol
Melting Pt: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD11100854
CAS Number: 10305-73-4
UN: 3077
ADR: 9+EHS,III