6-Aminocaproic acid 97%

Supplier: Apollo Scientific
Warning

APOSOR46222-1KGEA 148 EUR
APOSOR46222-1KG
6-Aminocaproic acid 97%
6-Aminocaproic acid
Formula: C₆H₁₃NO₂
MW: 131,17 g/mol
Melting Pt: 210…212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008238
EINECS: 200-469-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR