Methyl-2,4-dihydroxybenzoate
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Dihydroxybenzoic acid methyl ester, Methyl 2,4-dihydroxybenzoate
OR46230-100G OR46230-25G OR46230-500G
APOSOR46230-100GEA 72.5 EUR
APOSOR46230-100G APOSOR46230-25G APOSOR46230-500G
Methyl-2,4-dihydroxybenzoate
Methyl 2,4-dihydroxybenzoate
Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 116…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00002276
CAS Number: 2150-47-2