(3R)-(+)-3-(Dimethylamino)pyrrolidine 97%
Supplier: Apollo Scientific
Synonyms: (R)-(+)-3-(Dimethylamino)pyrrolidine, (R)-N,N-Dimethylpyrrolidin-3-amine
OR4630-5G OR4630-1G
APOSOR4630-5GEA 333 EUR
APOSOR4630-5G APOSOR4630-1G
(3R)-(+)-3-(Dimethylamino)pyrrolidine 97%
(3R)-(+)-3-(Dimethylamino)pyrrolidine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 166 °C
Density: 0.899 g/cm³ (25 °C)
Flash Pt: 51.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00191347
CAS Number: 132958-72-6
UN: 2734
ADR: 8+3,II