(R)-(+)-1-Boc-3-aminopyrrolidine 95%
Supplier: Apollo Scientific
Danger
Synonyms: (R)-1-Boc-3-aminopyrrolidine, (3R)-(+)-1-Boc-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-Boc-pyrrolidine, (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate, (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine
OR4635-25G OR4635-5G
APOSOR4635-25GEA 71.2 EUR
APOSOR4635-25G APOSOR4635-5G
(R)-(+)-1-Boc-3-aminopyrrolidine 95%
(R)-(+)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
Boiling Pt: 243…244 °C
Density: 1.098 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD03419272
CAS Number: 147081-49-0
UN: 2810
ADR: 6.1,III