(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine
Supplier: Apollo Scientific
Warning
Synonyms: (S)-tert-Butyl pyrrolidin-3-ylcarbamate, (3S)-(-)-3-(Boc-amino)pyrrolidine
OR4636-25G OR4636-5G OR4636-100G
APOSOR4636-25GEA 88.6 EUR
APOSOR4636-25G APOSOR4636-5G APOSOR4636-100G
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine
Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00143194
CAS Number: 122536-76-9