4-Perhydroazepinone hydrochloride
Supplier: Apollo Scientific
Danger
Synonyms: 4-Azepanone hydrochloride, Hexahydro-4-azepinone hydrochloride
OR4647-1G OR4647-5G
APOSOR4647-1GEA 21.9 EUR
APOSOR4647-1G APOSOR4647-5G
4-Perhydroazepinone hydrochloride
4-Perhydroazepinone hydrochloride
Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
Melting Pt: 176…181 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02183487
CAS Number: 50492-22-3