Diphenyl ether
Supplier: Apollo Scientific
Danger
OR46509-100G OR46509-1KG OR46509-500G
APOSOR46509-100GEA 21.9 EUR
APOSOR46509-100G APOSOR46509-1KG APOSOR46509-500G
Diphenyl ether
Diphenyl ether
Formula: C₁₂H₁₀O
Boiling Pt: 258…260 °C
Melting Pt: 25…27 °C
Density: 1.073 g/cm³ (25 °C)
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00003034
CAS Number: 101-84-8
EINECS: 202-981-2
UN: 3077
ADR: 9+EHS,III