1,5-Dibromo-2,4-dimethylbenzene
Supplier: Apollo Scientific
Warning
Synonyms: 4,6-Dibromo-m-xylene
OR46516-10G OR46516-25G OR46516-5G
APOSOR46516-10GEA 59.6 EUR
APOSOR46516-10G APOSOR46516-25G APOSOR46516-5G
1,5-Dibromo-2,4-dimethylbenzene
1,5-Dibromo-2,4-dimethylbenzene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Melting Pt: 71…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00154984
CAS Number: 615-87-2