3-Bromothioanisole
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromophenyl methyl sulphide, 1-Bromo-3-(methylthio)benzene
OR4652-100G OR4652-25G
APOSOR4652-100GEA 161 EUR
APOSOR4652-100G APOSOR4652-25G
3-Bromothioanisole
3-Bromothioanisole
Formula: C₇H₇BrS
MW: 203.1 g/mol
Boiling Pt: 124…125 °C/10mmHg
Density: 1.51 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041395
CAS Number: 33733-73-2
UN: 3334
ADR: 9,III