1-Methylbenzimidazole 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Methyl-1H-benzimidazole
OR46520-10G OR46520-25G
APOSOR46520-10GEA 23.2 EUR
APOSOR46520-10G APOSOR46520-25G
1-Methylbenzimidazole 98%
1-Methylbenzimidazole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 154 °C/12mmHg
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00192275
CAS Number: 1632-83-3