Methyl-2,5-dichloronicotinate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2,5-dichloronicotinate
OR46527-1G OR46527-500MG
APOSOR46527-1GEA 29 EUR
APOSOR46527-1G APOSOR46527-500MG
Methyl-2,5-dichloronicotinate
Methyl 2,5-dichloronicotinate
Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD03844857
CAS Number: 67754-03-4