5-Amino-2-methoxypyridine 97%
Supplier: Apollo Scientific
Warning
OR4654-25G OR4654-100G
APOSOR4654-25GEA 92.4 EUR
APOSOR4654-25G APOSOR4654-100G
5-Amino-2-methoxypyridine 97%
5-Amino-2-methoxypyridine
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 85…90 °C/1mmHg
Melting Pt: 29…31 °C
Density: 1.575 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006264
CAS Number: 6628-77-9
EINECS: 229-612-8