2-Bromo-5-iodoaniline 96%
Supplier: Apollo Scientific
Warning
OR46544-1G OR46544-250MG OR46544-5G
APOSOR46544-1GEA 72.5 EUR
APOSOR46544-1G APOSOR46544-250MG APOSOR46544-5G
2-Bromo-5-iodoaniline 96%
2-Bromo-5-iodoaniline
Formula: C₆H₅BrIN
Melting Pt: 70…75 °C
Storage Temperature: Ambient
MDL Number: MFCD16038675
CAS Number: 111721-74-5