Dichloro(1,5-cyclooctadiene)palladium(II) 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,5-Cyclooctadienepalladium(II) Dichloride
OR46568-1G OR46568-5G
APOSOR46568-1GEA 108 EUR
APOSOR46568-1G APOSOR46568-5G
Dichloro(1,5-cyclooctadiene)palladium(II) 98%
Dichloro(1,5-cyclooctadiene)palladium(II)
Formula: C₈H₁₂Cl₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00012412
CAS Number: 12107-56-1
EINECS: 235-161-8