4-Chloro-1H-imidazole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-1H-imidazole
OR46577-1G OR46577-5G
APOSOR46577-1GEA 62.5 EUR
APOSOR46577-1G APOSOR46577-5G
4-Chloro-1H-imidazole 98%
4-Chloro-1H-imidazole
Formula: C₃H₃ClN₂
MW: 102.52 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09033087
CAS Number: 15965-31-8