2-(Methylthio)benzoyl chloride
Supplier: Apollo Scientific
OR46685-1G OR46685-250MG OR46685-500MG
APOSOR46685-1GEA 418 EUR
APOSOR46685-1G APOSOR46685-250MG APOSOR46685-500MG
2-(Methylthio)benzoyl chloride
2-(Methylthio)benzoyl chloride
Formula: C₈H₇ClOS
MW: 186.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652997
CAS Number: 1442-03-1