Tetrabutylammonium hydrogen sulphate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate
OR4670-1KG OR4670-500G OR4670-5KG
APOSOR4670-1KGEA 154 EUR
APOSOR4670-1KG APOSOR4670-500G APOSOR4670-5KG
Tetrabutylammonium hydrogen sulphate 97%
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
MW: 339.54 g/mol
Melting Pt: 168…172 °C
Density: 1.01 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5