3-Methoxypyridine
Supplier: Apollo Scientific
Warning
OR4691-1G OR4691-5G OR4691-25G
APOSOR4691-1GEA 21.9 EUR
APOSOR4691-1G APOSOR4691-5G APOSOR4691-25G
3-Methoxypyridine
3-Methoxypyridine
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 65 °C/15mmHg
Density: 1.083 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00673022
CAS Number: 7295-76-3