2'-Methylacetanilide 98+%
Supplier: Apollo Scientific
Warning
OR4696-1KG OR4696-250G OR4696-25G
APOSOR4696-1KGEA 743 EUR
APOSOR4696-1KG APOSOR4696-250G APOSOR4696-25G
2'-Methylacetanilide 98+%
2'-Methylacetanilide
Formula: C₉H₁₁NO
Boiling Pt: 296 °C
Melting Pt: 109…110 °C
Density: 1.16 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014961
CAS Number: 120-66-1
EINECS: 204-414-4