N,N-Dimethyl-1-(1H-pyrrol-2-yl)methanamine
Supplier: Apollo Scientific
Synonyms: 2-[(Dimethylamino)methyl]pyrrole
OR470434-1G OR470434-5G
APOSOR470434-1GEA 256 EUR
APOSOR470434-1G APOSOR470434-5G
N,N-Dimethyl-1-(1H-pyrrol-2-yl)methanamine
N,N-Dimethyl-1-(1H-pyrrol-2-yl)methanamine
Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12028061
CAS Number: 14745-84-7