4-Chloro-2,6-dinitrobenzonitrile
Supplier: Apollo Scientific
OR471101-1G OR471101-5G
APOSOR471101-1GEA 448 EUR
APOSOR471101-1G APOSOR471101-5G
4-Chloro-2,6-dinitrobenzonitrile
4-Chloro-2,6-dinitrobenzonitrile
Formula: C₇H₂ClN₃O₄
Storage Temperature: Ambient
MDL Number: MFCD18651708
CAS Number: 1272756-26-9