1-Bromo-2-methyl-3,4-dinitrobenzene
Supplier: Apollo Scientific
OR471504-1G OR471504-5G
APOSOR471504-1GEA 107 EUR
APOSOR471504-1G APOSOR471504-5G
1-Bromo-2-methyl-3,4-dinitrobenzene
1-Bromo-2-methyl-3,4-dinitrobenzene
Formula: C₇H₅BrN₂O₄
MW: 261.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18642363
CAS Number: 290353-57-0