(-)-Bis[(S)-1-phenylethyl]amine
Supplier: Apollo Scientific
Warning
Synonyms: Bis[(S)-1-phenylethyl]amine
OR471545-5G OR471545-25G
APOSOR471545-5GEA 104 EUR
APOSOR471545-5G APOSOR471545-25G
(-)-Bis[(S)-1-phenylethyl]amine
(-)-Bis[(S)-1-phenylethyl]amine
Formula: C₁₆H₁₉N
MW: 225.34 g/mol
Boiling Pt: 98 °C/0.3mmHg
Density: 0.987 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00243087
CAS Number: 56210-72-1