2-sulphanyl-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-ol
Supplier: Apollo Scientific
Warning
Synonyms: 2-Mercapto-6,7-dihydro-3H-cyclopentapyrimidin-4(5H)-one
OR471549-1G OR471549-5G
APOSOR471549-1GEA 286 EUR
APOSOR471549-1G APOSOR471549-5G
2-sulphanyl-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-ol
2-sulphanyl-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-ol
Formula: C₇H₈N₂OS
MW: 168.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00223637
CAS Number: 35563-27-0