2-Bromo-4,6-xylidine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Bromo-2,4-dimethylaniline, 6-Bromo-2,4-xylidine, 2-Bromo-4,6-dimethylaniline
OR4720-25G
OR4720-100G
APOSOR4720-25GEA
63.8
EUR
APOSOR4720-25G
APOSOR4720-100G
2-Bromo-4,6-xylidine
2-Bromo-4,6-xylidine
Formula:
C₈H₁₀BrN MW: 200.08 g/mol Boiling Pt: 75…79 °C/5mmHg Melting Pt: 42…47 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047826 CAS Number: 41825-73-4 EINECS: 246-337-9 |