5,6-Dichloropyridine-3-sulphonyl chloride
Supplier: Apollo Scientific
Danger
OR4733-1G OR4733-250MG
APOSOR4733-1GEA 847 EUR
APOSOR4733-1G APOSOR4733-250MG
5,6-Dichloropyridine-3-sulphonyl chloride
5,6-Dichloropyridine-3-sulphonyl chloride
Formula: C₅H₂Cl₃NO₂S
MW: 246.5 g/mol
Melting Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD03541048
CAS Number: 98121-40-5
UN: 1759
ADR: 8,III