Allene 96%
Supplier: Apollo Scientific
Danger
Synonyms: Propa-1,2-diene
OR4738-25G OR4738-100G
APOSOR4738-25GEA 403 EUR
APOSOR4738-25G APOSOR4738-100G
Allene 96%
Allene
Contains up to 4% propene.
Formula: C₃H₄
Boiling Pt: –34 °C
Melting Pt: –136 °C
Density: 0.647 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008566
CAS Number: 463-49-0
EINECS: 207-335-3
UN: 2200
ADR: 2.1,