4-Chloro-3-nitrotoluene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-4-methyl-2-nitrobenzene
OR4743-500G OR4743-100G
APOSOR4743-500GEA 419 EUR
APOSOR4743-500G APOSOR4743-100G
4-Chloro-3-nitrotoluene 97%
4-Chloro-3-nitrotoluene
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 260…262 °C
Melting Pt: 7 °C
Density: 1.297 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007085
CAS Number: 89-60-1
EINECS: 201-922-8
UN: 2433
ADR: 6.1 III