2-Chloro-4-nitropyridine
Supplier: Apollo Scientific
Warning
OR4748-25G OR4748-5G
APOSOR4748-25GEA 40.6 EUR
APOSOR4748-25G APOSOR4748-5G
2-Chloro-4-nitropyridine
2-Chloro-4-nitropyridine
Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 50…56 °C
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00661454
CAS Number: 23056-36-2