3-Pyridone
Supplier: Apollo Scientific
Warning
Synonyms: 3-Hydroxypyridine, β-Pyridone, Pyridin-3-ol, 3-Pyridinol
OR4752-100G OR4752-1KG OR4752-500G
APOSOR4752-100GEA 53.8 EUR
APOSOR4752-100G APOSOR4752-1KG APOSOR4752-500G
3-Pyridone
3-Pyridone
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 180 °C/22mmHg
Melting Pt: 99…100 °C
Density: 1.37 g/cm³ (25 °C)
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00006378
CAS Number: 109-00-2
EINECS: 203-637-4