2-Amino-6-chlorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
OR4768-1G OR4768-500MG
APOSOR4768-1GEA 24.8 EUR
APOSOR4768-1G APOSOR4768-500MG
2-Amino-6-chlorobenzonitrile 98%
2-Amino-6-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00015546
CAS Number: 6575-11-7
EINECS: 229-501-4
UN: 3439
ADR: 6.1,III