2,4,6-Trichlorobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4,6-Trichlorobenzoyl Chloride
OR47821-25G OR47821-5G
APOSOR47821-25GEA 263 EUR
APOSOR47821-25G APOSOR47821-5G
2,4,6-Trichlorobenzoyl chloride 98%
2,4,6-Trichlorobenzoyl chloride
Formula: C₇H₂Cl₄O
MW: 243.9 g/mol
Boiling Pt: 107…108 °C/6mmHg
Density: 1.561 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075323
CAS Number: 4136-95-2
UN: 3265
ADR: 8,II