1,2,4,5-Tetrabromobenzene
Supplier: Apollo Scientific
Warning
OR47827-1G OR47827-5G
APOSOR47827-1GEA 21.9 EUR
APOSOR47827-1G APOSOR47827-5G
1,2,4,5-Tetrabromobenzene
1,2,4,5-Tetrabromobenzene
Formula: C₆H₂Br₄
MW: 393.7 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00000063
CAS Number: 636-28-2
EINECS: 211-253-3