Trichloroacetonitrile 99%
Supplier: Apollo Scientific
|
Danger
|
OR4786-1KG
OR4786-250G
OR4786-5KG
APOSOR4786-1KGEA
174
EUR
APOSOR4786-1KG
APOSOR4786-250G
APOSOR4786-5KG
Trichloroacetonitrile 99%
Trichloroacetonitrile
Formula:
C₂Cl₃N MW: 144.39 g/mol Boiling Pt: 83…84 °C Melting Pt: –42 °C Density: 1.44 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001842 CAS Number: 545-06-2 EINECS: 208-885-7 UN: 3276 ADR: 6.1 III Merck Index: 12,09757 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...