(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetonitrile 95%
Supplier: Apollo Scientific
Synonyms: 2-(1,3-Dioxoisoindolin-2-yl)acetonitrile
OR480201-1G OR480201-500MG OR480201-5G
APOSOR480201-1GEA 210 EUR
APOSOR480201-1G APOSOR480201-500MG APOSOR480201-5G
(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetonitrile 95%
(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetonitrile
Formula: C₁₀H₆N₂O₂
MW: 186.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00087362
CAS Number: 3842-20-4