1-Methylimidazolidin-2-one 95%
Supplier: Apollo Scientific
Synonyms: 1-Methyl-2-imidazolidinone
OR480211-1G OR480211-2G OR480211-5G
APOSOR480211-1GEA 141 EUR
APOSOR480211-1G APOSOR480211-2G APOSOR480211-5G
1-Methylimidazolidin-2-one 95%
1-Methylimidazolidin-2-one
Formula: C₄H₈N₂O
MW: 100.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800601
CAS Number: 694-32-6