1-(2-Nitrophenyl)-1H-pyrazole 97%
Supplier: Apollo Scientific
OR480243-10G OR480243-1G OR480243-5G
APOSOR480243-10GEA 674 EUR
APOSOR480243-10G APOSOR480243-1G APOSOR480243-5G
1-(2-Nitrophenyl)-1H-pyrazole 97%
1-(2-Nitrophenyl)-1H-pyrazole
Formula: C₉H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06661759
CAS Number: 25688-17-9