4-Chloro-2,5-xylidine 98%
Supplier: Apollo Scientific
Synonyms: 4-Chloro-2,5-dimethylaniline
OR480249-100G OR480249-25G
APOSOR480249-100GEA 1180 EUR
APOSOR480249-100G APOSOR480249-25G
4-Chloro-2,5-xylidine 98%
4-Chloro-2,5-xylidine
Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08445705
CAS Number: 20782-94-9